Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloropyrimidine-5-boronic acid, 96%

Click to view available options
Quantity:
250mg
1g
5g
1003845-06-4
C4H4BClN2O2
158.348
MFCD08063113
YTCIHPTZKKWKKC-UHFFFAOYSA-N
2-chloropyrimidine-5-boronic acid,2-chloro-5-pyrimidineboronic acid,2-chloropyrimidin-5-yl boronic acid,acmc-2097pq,ksc493q8p,2-chloropyrimidin-5-boronic acid,2-chloro-5-pyrimidinylboronic acid,2-chloro-pyridineyl-5-boronic acid,2-chloro-pyrimidine-5-boronic acid,2-chloropyrimidin-5-yl boronic acid
21747362
(2-chloropyrimidin-5-yl)boronic acid
B(C1=CN=C(N=C1)Cl)(O)O
1003845-06-4
C4H4BClN2O2
158.348
MFCD08063113
YTCIHPTZKKWKKC-UHFFFAOYSA-N
2-chloropyrimidine-5-boronic acid,2-chloro-5-pyrimidineboronic acid,2-chloropyrimidin-5-yl boronic acid,acmc-2097pq,ksc493q8p,2-chloropyrimidin-5-boronic acid,2-chloro-5-pyrimidinylboronic acid,2-chloro-pyridineyl-5-boronic acid,2-chloro-pyrimidine-5-boronic acid,2-chloropyrimidin-5-yl boronic acid
21747362
(2-chloropyrimidin-5-yl)boronic acid
B(C1=CN=C(N=C1)Cl)(O)O
Quantity 5g
Color White
Formula Weight 158.35
Physical Form Powder
Percent Purity 96%
Chemical Name or Material 2-Chloropyrimidine-5-boronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C