Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloropyrimidine-5-boronic acid pinacol ester, 95%, Acros Organics

Click to view available options
Quantity:
1g
1003845-08-6
C10H14BClN2O2
240.5
VLAPDEKXZLRRKV-UHFFFAOYSA-N
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,2-chloropyrimidine-5-boronic acid pinacol ester,2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,2-chloropyrimidin-5-ylboronic acid pinacol ester,2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidine,2-chloropyrimidine-5-boronic acid, pinacol ester,2-chloropyrimidin-5-yl boronic acid pinacol ester,2-5-chloro-2-pyrimidyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-chloranyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,pyrimidine, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
51341943
2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)Cl
1003845-08-6
C10H14BClN2O2
240.5
VLAPDEKXZLRRKV-UHFFFAOYSA-N
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,2-chloropyrimidine-5-boronic acid pinacol ester,2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,2-chloropyrimidin-5-ylboronic acid pinacol ester,2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidine,2-chloropyrimidine-5-boronic acid, pinacol ester,2-chloropyrimidin-5-yl boronic acid pinacol ester,2-5-chloro-2-pyrimidyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-chloranyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine,pyrimidine, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
51341943
2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)Cl
Packaging Glass Bottle
Melting Point 67.0°C to 69.0°C
Quantity 1g
Assay Percent Range 94% min. (GC)
Formula Weight 240.5
Percent Purity 95%
Chemical Name or Material 2-Chloropyrimidine-5-boronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning