Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloropyrimidine-5-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
374068-01-6
C5H3ClN2O2
158.54
DUCXUPKLVVSJKA-UHFFFAOYSA-N
2-chloro-pyrimidine-5-carboxylic acid,2-chloro-5-carboxypyrimidine,5-carboxy-2-chloropyrimidine,5-pyrimidinecarboxylicacid, 2-chloro,2-chloro-5-pyrimidinecarboxylic acid,5-pyrimidinecarboxylic acid, 2-chloro,2-chloropyrimidine-5-carboxylicacid,acmc-1aig3,2-chloropyrimidine 5-carboxylic acid,2-chloro-5-pyrimidine carboxylic acid
11094906
2-chloropyrimidine-5-carboxylic acid
C1=C(C=NC(=N1)Cl)C(=O)O
374068-01-6
C5H3ClN2O2
158.54
DUCXUPKLVVSJKA-UHFFFAOYSA-N
2-chloro-pyrimidine-5-carboxylic acid,2-chloro-5-carboxypyrimidine,5-carboxy-2-chloropyrimidine,5-pyrimidinecarboxylicacid, 2-chloro,2-chloro-5-pyrimidinecarboxylic acid,5-pyrimidinecarboxylic acid, 2-chloro,2-chloropyrimidine-5-carboxylicacid,acmc-1aig3,2-chloropyrimidine 5-carboxylic acid,2-chloro-5-pyrimidine carboxylic acid
11094906
2-chloropyrimidine-5-carboxylic acid
C1=C(C=NC(=N1)Cl)C(=O)O
Color White
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Melting Point 125.0°C to 127.5°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 158.54
Physical Form Powder or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Chloropyrimidine-5-carboxylic acid
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning