Alfa Aesar™ 2-Chloroquinoline-3-carboxaldehyde, 98%

Manufacturer: Alfa Aesar™ H2713303

Quantity 1g

 View more versions of this product

Catalog No. AAH2713303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 73568-25-9
    Molecular Formula C10H6ClNO
    Formula Weight 191.62
    MDL Number MFCD00130079
    Synonym 2-Chloro-3-formylquinoline
    Chemical Name or Material 2-Chloroquinoline-3-carboxaldehyde

    Specifications

    Formula Weight 191.62
    MDL Number MFCD00130079
    Health Hazard 1 Warning
    Melting Point 148° to 150°C
    Quantity 1g
    Molecular Formula C10H6ClNO
    CAS 73568-25-9
    Synonym 2-Chloro-3-formylquinoline
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338-P337+P313
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
    Chemical Name or Material 2-Chloroquinoline-3-carboxaldehyde
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338-P337+P313
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit