Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloroquinoline, 99%

Click to view available options
Quantity:
5g
25g
612-62-4
C9H6ClN
163.604
MFCD00006741
OFUFXTHGZWIDDB-UHFFFAOYSA-N
2-chloro-quinoline,quinoline, 2-chloro,chloroquinoline,unii-ux7rin8gew,quinoline, chloro,ccris 3977,ux7rin8gew,chioroquinoline,2-chloro quinoline,pubchem7580
11928
2-chloroquinoline
C1=CC=C2C(=C1)C=CC(=N2)Cl
612-62-4
C9H6ClN
163.604
MFCD00006741
OFUFXTHGZWIDDB-UHFFFAOYSA-N
2-chloro-quinoline,quinoline, 2-chloro,chloroquinoline,unii-ux7rin8gew,quinoline, chloro,ccris 3977,ux7rin8gew,chioroquinoline,2-chloro quinoline,pubchem7580
11928
2-chloroquinoline
C1=CC=C2C(=C1)C=CC(=N2)Cl
Applications
2-Chloroquinoline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It possess antimicrobial activity against a panel of bacterial and fungal strains. It reacts with benzotriazole, followed by cyclization with loss of nitrogen, gives the indolo[2,3-b]quinoline ring system for use in novel cytotoxic DNA topoisomerase II inhibitors.

Solubility
Soluble in ethanol, ether, methanol, and chloroform. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
Melting Point 34°C to 37°C
Odor Characteristic
Quantity 25g
Flash Point >110°C (230°F)
Boiling Point 265°C to 267°C
Beilstein 112561
Formula Weight 163.61
Percent Purity 99%
Chemical Name or Material 2-Chloroquinoline

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338-P337+P313
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : VB2320000

TSCA : TSCA