Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chlorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Click to view available options
Quantity:
5g
25g
100g
2039-87-4
C8H7Cl
138.594
MFCD00000567
ISRGONDNXBCDBM-UHFFFAOYSA-N
2-chlorostyrene,1-chloro-2-vinylbenzene,o-chlorostyrene,benzene, 1-chloro-2-ethenyl,styrene, o-chloro,ortho-chlorostyrene,stryene, o-chloro,styrene, 2-chloro,unii-za43r4q315,ccris 4598
14906
1-chloro-2-ethenylbenzene
C=CC1=CC=CC=C1Cl
2039-87-4
C8H7Cl
138.594
MFCD00000567
ISRGONDNXBCDBM-UHFFFAOYSA-N
2-chlorostyrene,1-chloro-2-vinylbenzene,o-chlorostyrene,benzene, 1-chloro-2-ethenyl,styrene, o-chloro,ortho-chlorostyrene,stryene, o-chloro,styrene, 2-chloro,unii-za43r4q315,ccris 4598
14906
1-chloro-2-ethenylbenzene
C=CC1=CC=CC=C1Cl
Applications
2-Chlorostyrene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Quantity 100g
Flash Point 65°C (149°F)
Melting Point -63°C
Odor Pungent
Boiling Point 189°C
Beilstein 2038863
Formula Weight 138.6
Percent Purity 98%
Chemical Name or Material 2-Chlorostyrene, stabilized with 0.1% 4-tert-butylcatechol

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : WL4160000

Recommended Storage : 2° to 8°C