Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chlorothiazole-4-carboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
5198-87-8
C4H2ClNO2S
163.58
UVYJJJQMZPCYKY-UHFFFAOYSA-N
2-chlorothiazole-4-carboxylic acid,2-chloro-4-thiazolecarboxylic acid,2-chloro-thiazole-4-carboxylic acid,4-carboxy-2-chloro-1,3-thiazole,4-thiazolecarboxylic acid, 2-chloro,2chloro-4-thiazolecarboxylic acid
21803026
2-chloro-1,3-thiazole-4-carboxylic acid
C1=C(N=C(S1)Cl)C(=O)O
5198-87-8
C4H2ClNO2S
163.58
UVYJJJQMZPCYKY-UHFFFAOYSA-N
2-chlorothiazole-4-carboxylic acid,2-chloro-4-thiazolecarboxylic acid,2-chloro-thiazole-4-carboxylic acid,4-carboxy-2-chloro-1,3-thiazole,4-thiazolecarboxylic acid, 2-chloro,2chloro-4-thiazolecarboxylic acid
21803026
2-chloro-1,3-thiazole-4-carboxylic acid
C1=C(N=C(S1)Cl)C(=O)O
Packaging Glass bottle
Color White to Yellow
Quantity 1g
Infrared Spectrum Authentic
Melting Point 220.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 163.58
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-chlorothiazole-4-carboxylic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning