Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorotoluene, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
500mL
1L
2.5L
Packaging:
Glass bottle
95-49-8
C7H7Cl
126.59
MFCD00000562
IBSQPLPBRSHTTG-UHFFFAOYSA-N
2-chlorotoluene,o-chlorotoluene,benzene, 1-chloro-2-methyl,o-tolyl chloride,ortho-chlorotoluene,toluene, o-chloro,2-methylchlorobenzene,halso 99,2-chloro-1-methylbenzene,1-methyl-2-chlorobenzene
7238
1-chloro-2-methylbenzene
CC1=CC=CC=C1Cl
95-49-8
C7H7Cl
126.59
MFCD00000562
IBSQPLPBRSHTTG-UHFFFAOYSA-N
2-chlorotoluene,o-chlorotoluene,benzene, 1-chloro-2-methyl,o-tolyl chloride,ortho-chlorotoluene,toluene, o-chloro,2-methylchlorobenzene,halso 99,2-chloro-1-methylbenzene,1-methyl-2-chlorobenzene
7238
1-chloro-2-methylbenzene
CC1=CC=CC=C1Cl
Specific Gravity 1.08
Color Undesignated
Density 1.0800g/mL
Quantity 1L
Infrared Spectrum Authentic
Water 0.015% max.
Refractive Index 1.5240 to 1.5260
Flash Point 49°C
Packaging Glass bottle
Boiling Point 157.0°C to 159.0°C
Melting Point -36.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4Cl
Beilstein 05, 290
Merck Index 15, 2175
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol, benzene, chlroform and, ether
Formula Weight 126.59
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Chlorotoluene
Show More Show Less

GHS P Statement
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Suspected of damaging the unborn child.

GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Obtain special instructions before use. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning