Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chlorotoluene, 99%

Click to view available options
Quantity:
100g
250g
1000g
95-49-8
C7H7Cl
126.583
MFCD00000562
IBSQPLPBRSHTTG-UHFFFAOYSA-N
2-chlorotoluene,o-chlorotoluene,benzene, 1-chloro-2-methyl,o-tolyl chloride,ortho-chlorotoluene,toluene, o-chloro,2-methylchlorobenzene,halso 99,2-chloro-1-methylbenzene,1-methyl-2-chlorobenzene
7238
1-chloro-2-methylbenzene
CC1=CC=CC=C1Cl
95-49-8
C7H7Cl
126.583
MFCD00000562
IBSQPLPBRSHTTG-UHFFFAOYSA-N
2-chlorotoluene,o-chlorotoluene,benzene, 1-chloro-2-methyl,o-tolyl chloride,ortho-chlorotoluene,toluene, o-chloro,2-methylchlorobenzene,halso 99,2-chloro-1-methylbenzene,1-methyl-2-chlorobenzene
7238
1-chloro-2-methylbenzene
CC1=CC=CC=C1Cl
Applications
2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Melting Point -36°C
Flash Point 47°C (116°F)
Quantity 250g
Boiling Point 157°C to 159°C
UN Number UN2238
Beilstein 1904175
Merck Index 14,2171
Formula Weight 126.59
Percent Purity 99%
Chemical Name or Material 2-Chlorotoluene

GHS H Statement
H226-H332
Flammable liquid and vapour.
Harmful if inhaled.

GHS P Statement
P210-P261-P280-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROTOLUENES

RTECSNumber : XS9000000

TSCA : TSCA