Alfa Aesar™ 2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 100-200 mesh, 1.0-1.4 mmol/g

Manufacturer: Alfa Aesar™ 4440709

Quantity 10g

 View more versions of this product

Catalog No. AA4440709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 42074-68-0
    MDL Number MFCD00040399
    Chemical Name or Material 2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 1.0 to 1.4 mmol/g

    Specifications

    Physical Form Powder
    CAS 42074-68-0
    MDL Number MFCD00040399
    Mesh Size 100 to 200 Mesh
    Quantity 10g
    Chemical Name or Material 2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 1.0 to 1.4 mmol/g

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit