Learn More
2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 100-200 mesh, 1.0-1.4 mmol/g, Thermo Scientific Chemicals
Description
2-Chlorotrityl chloride is used as an intermediate of clotrimazole. It is also used in convergent peptide synthesis. Furthermore, it is used in the preparation of modified trityl nucleosides. In addition, it serves as a protecting group reagent for carboxylic acids forming esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 50 g |
Mesh Size | 100 to 200 Mesh |
Chemical Name or Material | 2-Chlorotrityl chloride on polystyrene, 1% cross-linked, 1.0 to 1.4 mmol/g |
Safety and Handling
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.