Alfa Aesar™ 2-Chloroveratraldehyde, 98%

Manufacturer: Alfa Aesar™ A1062306

Quantity 5g

 View more versions of this product

Catalog No. AAA1062306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5417-17-4
    Molecular Formula C9H9ClO3
    Formula Weight 200.62
    MDL Number MFCD00084942
    Beilstein 2109344
    Synonym 2-Chloro-3,4-dimethoxybenzaldehyde
    Chemical Name or Material 2-Chloroveratraldehyde

    Specifications

    Formula Weight 200.62
    Molecular Formula C9H9ClO3
    CAS 5417-17-4
    Beilstein 2109344
    Synonym 2-Chloro-3,4-dimethoxybenzaldehyde
    MDL Number MFCD00084942
    Melting Point 70° to 72°C
    Quantity 5g
    Chemical Name or Material 2-Chloroveratraldehyde
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit