Learn More
Tocris Bioscience™ 2-CMDO Available on GSA/VA Contract for Federal Government customers only.
D2-like antagonist; Displays some D4 selectivity
Manufacturer:  Tocris Bioscience™ 0782/10
 View more versions of this product
Catalog No. 07-821-0
Add to cart
Description
Description
Dopamine D2-like receptor antagonist that displays some selectivity for D4 receptors (Ki values are 0.54 and 2.5nM for D4 and D2 receptors respectively).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
2-Chloro-11-(4-methylpiperazino)dibenz(Z)[b,f]oxepin maleate
C19H19ClN2O.C4H4O4
10mg
24140-98-5
Observed MW: 442.9
Soluble to 100mM in DMSO
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT