Learn More
Tocris Bioscience™ 2-CMDO Available on GSA/VA Contract for Federal Government customers only.
D2-like antagonist; displays some D4 selectivity
$149.00 - $619.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
07-825-0 View Documents Tocris Bioscience™
0782/50
Each for $619.00
Add to cart
07-821-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
0782/10
Each for $149.00
Add to cart
Description
Description
Dopamine D2-like receptor antagonist that displays some selectivity for D4 receptors (Ki values are 0.54 and 2.5 nM for D4 and D2 receptors respectively).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.