Learn More
Tocris Bioscience™ 2-CMDO
D2-like antagonist; Displays some D4 selectivity
Manufacturer:  Tocris Bioscience™ 0782/50
 View more versions of this product
Catalog No. 07-825-0
Add to cart
Description
Description
Dopamine D2-like receptor antagonist that displays some selectivity for D4 receptors (Ki values are 0.54 and 2.5nM for D4 and D2 receptors respectively).
Specifications
Specifications
2-Chloro-11-(4-methylpiperazino)dibenz(Z)[b,f]oxepin maleate
C19H19ClN2O.C4H4O4
50mg
24140-98-5
Observed MW: 442.9
Soluble to 100mM in DMSO
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT