Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPEX CertiPrep™ 2-Color Aspergillus Detection Kit

Provides a rapid and accurate test to determine the presence of Aspergillus DNA in a sample.

Supplier:  SPEX CertiPrep™ ASPKIT2500

 View more versions of this product

Catalog No. 01-184-144


Add to Cart

Alert:

Storage Conditions: -20°C; Shipping Conditions: Expanded Polystyrene (EPS) containers for thermal insulation and ice packs


Description

Description

  • The kit uses quantitative PCR (qPCR) to measure the total amount of the four most common strains of Aspergillus: niger, fumigatus, flavus and terrus
  • Probes for the four strains of the Aspergillus are labeled with the FAM dye, and an internal control is labeled with HEX/VIC
  • The internal control ensures the sample is free of PCR inhibitors that would lead to false negative results. To help distinguish the Master Mix from the template DNA and from the final product during sample preparation, an inert yellow dye is provided to label the template prior to adding to the Master Mix which contains an inert blue dye
  • When mixed the resulting solution is green, confirming the presence of all components
  • The kit works with any qPCR system that supports the two color channels
  • It is very sensitive and can detect down to 10 copies of Aspergillus DNA
Specifications

Specifications

Aspergillus detection MasterMix, internal control, positive control, 20X yellow dye, ROX, DNase/RNase/protease free water
qPCR
>95%
qPCR kit for test for pathogens
500
Liquid
Taq DNA Polymerase
Most quantitative PCR machines that are high, low or no ROX
Modification of the Plutheros Protocol
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit