Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Cyano-2,2-dimethylacetic acid, 95%

Click to view available options
Quantity:
250mg
1g
22426-30-8
C5H7NO2
113.116
MFCD11100210
DZDVNAVPCCNJRK-UHFFFAOYSA-N
2-cyano-2,2-dimethylacetic acid,2-cyano-2-methyl-propanoic acid,cyanomethylpropanoicacid,2-cyanoisobutyric acid,dimethylcyanoacetic acid,propanoic acid, 2-cyano-2-methyl
11367091
2-cyano-2-methylpropanoic acid
CC(C)(C#N)C(=O)O
22426-30-8
C5H7NO2
113.116
MFCD11100210
DZDVNAVPCCNJRK-UHFFFAOYSA-N
2-cyano-2,2-dimethylacetic acid,2-cyano-2-methyl-propanoic acid,cyanomethylpropanoicacid,2-cyanoisobutyric acid,dimethylcyanoacetic acid,propanoic acid, 2-cyano-2-methyl
11367091
2-cyano-2-methylpropanoic acid
CC(C)(C#N)C(=O)O
Boiling Point 132°C to 135°C (12mmHg)
Sensitivity Moisture Sensitive
Quantity 250mg
Color White
Melting Point 56°C to 61°C
UN Number UN2923
Formula Weight 113.11
Physical Form Crystalline Solid
Percent Purity 95%
Chemical Name or Material 2-Cyano-2,2-dimethylacetic acid

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.