Alfa Aesar™ 2-Cyano-5-fluoropyridine, 98+%

Manufacturer: Alfa Aesar™ H2595606

Quantity 5g

 View more versions of this product

Catalog No. AAH2595606

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 327056-62-2
    Molecular Formula C6H3FN2
    Formula Weight 122.1
    MDL Number MFCD08741371
    Synonym 5-Fluoropicolinonitrile; 5-Fluoropyridine-2-carbonitrile
    Chemical Name or Material 2-Cyano-5-fluoropyridine

    Specifications

    Formula Weight 122.1
    Molecular Formula C6H3FN2
    MDL Number MFCD08741371
    CAS 327056-62-2
    Synonym 5-Fluoropicolinonitrile; 5-Fluoropyridine-2-carbonitrile
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point 41° to 44°C
    Quantity 5g
    Chemical Name or Material 2-Cyano-5-fluoropyridine
    Assay ≥98%

    GHS H Statement
    H302-H315-H319
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit