Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Cyano-5-(trifluoromethyl)pyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
95727-86-9
C7H3F3N2
172.11
MFCD01001119
WDSCJULUXJSJOX-UHFFFAOYSA-N
5-trifluoromethyl picolinonitrile,5-trifluoromethyl pyridine-2-carbonitrile,2-cyano-5-trifluoromethyl pyridine,2-cyano-5-trifluoromethylpyridine,5-trifluoromethyl-2-pyridinecarbonitrile,5-trifluoromethyl-pyridine-2-carbonitrile,2-pyridinecarbonitrile, 5-trifluoromethyl,pubchem17518,5-trifluoromethyl ;picolinonitrile,2-cyano-5-trifluoromethyl pyridine, 5-trifluoromethyl picolinonitrile
1535372
5-(trifluoromethyl)pyridine-2-carbonitrile
C1=CC(=NC=C1C(F)(F)F)C#N
95727-86-9
C7H3F3N2
172.11
MFCD01001119
WDSCJULUXJSJOX-UHFFFAOYSA-N
5-trifluoromethyl picolinonitrile,5-trifluoromethyl pyridine-2-carbonitrile,2-cyano-5-trifluoromethyl pyridine,2-cyano-5-trifluoromethylpyridine,5-trifluoromethyl-2-pyridinecarbonitrile,5-trifluoromethyl-pyridine-2-carbonitrile,2-pyridinecarbonitrile, 5-trifluoromethyl,pubchem17518,5-trifluoromethyl ;picolinonitrile,2-cyano-5-trifluoromethyl pyridine, 5-trifluoromethyl picolinonitrile
1535372
5-(trifluoromethyl)pyridine-2-carbonitrile
C1=CC(=NC=C1C(F)(F)F)C#N
Applications
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 250mg
UN Number UN3439
Beilstein 13339826
Formula Weight 172.11
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Cyano-5-(trifluoromethyl)pyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.