Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyano-6-methylpyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-3771065
Click to view available options
Quantity:
1 g
5 g
1620-75-3
C7H6N2
118.14
MFCD00190586
CMADFEQMYFNYCF-UHFFFAOYSA-N
6-methylpicolinonitrile, 2-cyano-6-methylpyridine, 6-methyl-2-pyridinecarbonitrile, 2-methyl-6-cyanopyridine, 6-methyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 6-methyl, 6-methyl-2-cyanopyridine, 2-cyano-6-methyl pyridine, 2-cyano-6-methylpyridine 2-cyano-6-picoline, 6-methylpyridin-2-carbonitril
74182
6-methylpyridine-2-carbonitrile
CC1=CC=CC(=N1)C#N
1620-75-3
C7H6N2
118.14
MFCD00190586
CMADFEQMYFNYCF-UHFFFAOYSA-N
6-methylpicolinonitrile, 2-cyano-6-methylpyridine, 6-methyl-2-pyridinecarbonitrile, 2-methyl-6-cyanopyridine, 6-methyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 6-methyl, 6-methyl-2-cyanopyridine, 2-cyano-6-methyl pyridine, 2-cyano-6-methylpyridine 2-cyano-6-picoline, 6-methylpyridin-2-carbonitril
74182
6-methylpyridine-2-carbonitrile
CC1=CC=CC(=N1)C#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 70.0°C to 74.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 118.14
Percent Purity 97%
Chemical Name or Material 2-Cyano-6-methylpyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 216-588-9

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit