Alfa Aesar™ 2-Cyanobenzaldehyde, 98%

Manufacturer: Alfa Aesar™ H2902706

Quantity 5g

 View more versions of this product

Catalog No. AAH2902706

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7468-67-9
    Molecular Formula C8H5NO
    Formula Weight 131.13
    MDL Number MFCD00017503
    Synonym 2-Formylbenzonitrile
    Chemical Name or Material 2-Cyanobenzaldehyde

    Specifications

    Formula Weight 131.13
    Odor Characteristic
    Molecular Formula C8H5NO
    MDL Number MFCD00017503
    CAS 7468-67-9
    Synonym 2-Formylbenzonitrile
    Health Hazard 1 Warning
    Melting Point 102° to 106°C
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Quantity 5g
    Chemical Name or Material 2-Cyanobenzaldehyde
    Assay 98%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit