Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanobenzoic acid, 94%, Thermo Scientific Chemicals

Catalog Number AAH5023706
Click to view available options
Quantity:
1 g
5 g
3839-22-3
C8H5NO2
147.133
MFCD00045796
DTNSDCJFTHMDAK-UHFFFAOYSA-N
o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v
138061
2-cyanobenzoic acid
C1=CC=C(C(=C1)C#N)C(=O)O
3839-22-3
C8H5NO2
147.133
MFCD00045796
DTNSDCJFTHMDAK-UHFFFAOYSA-N
o-cyanobenzoic acid, cyanobenzoic acid, benzoic acid, 2-cyano, 2-carboxybenzonitrile, 2-carboxy-benzonitrile, 2-cyanobenzoicacid, 2-cyano-benzoic acid, pubchem12513, acmc-1bn2v
138061
2-cyanobenzoic acid
C1=CC=C(C(=C1)C#N)C(=O)O

2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyanobenzoic acid has been used as reactant in the synthesis of T-box riboswitch antiterminator RNA-binding oxazolidinone derivatives, Benzoate modified β-cyclodextrin derivatives and Pyrrolo[1,2-f]triazines for use as JAK2 inhibitors. As a reactant involved in decarboxylation, decarboxylative halogenation and protodecarboxylation.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Melting Point 210°C to 212°C
Quantity 5 g
Formula Weight 147.13
Percent Purity 96%
Chemical Name or Material 2-Cyanobenzoic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit