Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-cyanoethyl N,n-diisopropylchlorophosphoramidite, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
89992-70-1
C9H18ClN2OP
236.68
MFCD00011544
QWTBDIBOOIAZEF-UHFFFAOYSA-N
2-cyanoethyl n,n-diisopropylchlorophosphoramidite,2-cyanoethyl-n,n-diisopropylchlorophosphoramidite,phosphoramidochloridous acid, bis 1-methylethyl-, 2-cyanoethyl ester,2-cyanoethoxy diisopropylamino chlorophosphine,3-chloro diisopropylamino phosphino oxy propanenitrile,3-chloro-diisopropylamino phosphanyl oxypropanenitrile,3-chloro diisopropylamino phosphanyl oxy propanenitrile,acmc-20anxd,2-cyanoethyl diisopropylchloro-phosphoramidite,2-cyanoethyl diisopropylphosphoramidochloridite
2734844
3-[chloro-[di(propan-2-yl)amino]phosphanyl]oxypropanenitrile
CC(C)N(C(C)C)P(OCCC#N)Cl
89992-70-1
C9H18ClN2OP
236.68
MFCD00011544
QWTBDIBOOIAZEF-UHFFFAOYSA-N
2-cyanoethyl n,n-diisopropylchlorophosphoramidite,2-cyanoethyl-n,n-diisopropylchlorophosphoramidite,phosphoramidochloridous acid, bis 1-methylethyl-, 2-cyanoethyl ester,2-cyanoethoxy diisopropylamino chlorophosphine,3-chloro diisopropylamino phosphino oxy propanenitrile,3-chloro-diisopropylamino phosphanyl oxypropanenitrile,3-chloro diisopropylamino phosphanyl oxy propanenitrile,acmc-20anxd,2-cyanoethyl diisopropylchloro-phosphoramidite,2-cyanoethyl diisopropylphosphoramidochloridite
2734844
3-[chloro-[di(propan-2-yl)amino]phosphanyl]oxypropanenitrile
CC(C)N(C(C)C)P(OCCC#N)Cl
Chloride 14.5 to 15.5%
Specific Gravity 1.061
Infrared Spectrum Authentic
Density 1.0610g/mL
Refractive Index 1.4760 to 1.4800
Color White to Yellow
Flash Point 110°C
NMR 96.0% min.
Quantity 1g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula [(CH3)2CH]2NP(Cl)O(CH2)2CN
Solubility Information Solubility in water: reacts.
Formula Weight 236.68
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Cyanoethyl N,N-diisopropylchlorophosphoramidite
Show More Show Less

GHS H Statement
Catches fire spontaneously if exposed to air.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas.
Protect from moisture.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a PO

GHS Signal Word: Danger

Recommended Storage : May develop turbidity on storage