Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite, 95%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
102691-36-1
C15H32N3OP
301.41
RKVHNYJPIXOHRW-UHFFFAOYSA-N
3-bis diisopropylamino phosphino oxy propanenitrile,bis diisopropylamino 2-cyanoethoxy phosphine,2-cyanoethyl n,n,n',n'-tetraisopropylphosphorodiamidite,2-cyanoethyl-n,n,n',n'-tetraisopropylphosphorodiamidite,2-cyanoethyl n,n,n',n'-tetraisopropylphosphordiamidite,phosphorodiamidous acid, tetrakis 1-methylethyl-, 2-cyanoethyl ester,2-cyanoethyl n,n,n',n'-tetraisopropyl-phosphordiamidite,2-cyanoethyl tetraisopropylphosphorodiamidite,cetipd,3-bis diisopropylamino phosphinooxy propanenitrile
128153
3-bis[di(propan-2-yl)amino]phosphanyloxypropanenitrile
CC(C)N(C(C)C)P(N(C(C)C)C(C)C)OCCC#N
102691-36-1
C15H32N3OP
301.41
RKVHNYJPIXOHRW-UHFFFAOYSA-N
3-bis diisopropylamino phosphino oxy propanenitrile,bis diisopropylamino 2-cyanoethoxy phosphine,2-cyanoethyl n,n,n',n'-tetraisopropylphosphorodiamidite,2-cyanoethyl-n,n,n',n'-tetraisopropylphosphorodiamidite,2-cyanoethyl n,n,n',n'-tetraisopropylphosphordiamidite,phosphorodiamidous acid, tetrakis 1-methylethyl-, 2-cyanoethyl ester,2-cyanoethyl n,n,n',n'-tetraisopropyl-phosphordiamidite,2-cyanoethyl tetraisopropylphosphorodiamidite,cetipd,3-bis diisopropylamino phosphinooxy propanenitrile
128153
3-bis[di(propan-2-yl)amino]phosphanyloxypropanenitrile
CC(C)N(C(C)C)P(N(C(C)C)C(C)C)OCCC#N
Specific Gravity 0.949
Quantity 25g
Refractive Index 1.47
Density 0.9490g/mL
Packaging Glass bottle
Boiling Point 98.0°C to 100.0°C (5.0mmHg)
Flash Point 60°C
Assay Percent Range 95%
Formula Weight 301.41
Percent Purity 95%
Chemical Name or Material 2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphorodiamidite
Show More Show Less