Alfa Aesar™ 2-Cyanoethylhydrazine, 95%

Manufacturer: Alfa Aesar™ B2522706

Quantity 5g

 View more versions of this product

Catalog No. AAB2522706

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 353-07-1
    Molecular Formula C3H7N3
    Linear Formula NCCH2CH2NHNH2
    Formula Weight 85.11
    MDL Number MFCD00043620
    UN Number UN2927
    Beilstein 773834
    Synonym 3-Hydrazinopropionitrile
    Chemical Name or Material 2-Cyanoethylhydrazine

    Specifications

    Formula Weight 85.11
    Linear Formula NCCH2CH2NHNH2
    MDL Number MFCD00043620
    CAS 353-07-1
    UN Number UN2927
    Synonym 3-Hydrazinopropionitrile
    Health Hazard 1 Danger
    Boiling Point 76° to 79°C (0.5mmHg)
    Health Hazard 2 GHS H Statement
    H300-H314
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
    Flash Point 109°C (228°F)
    Quantity 5g
    Molecular Formula C3H7N3
    Beilstein 773834
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Chemical Name or Material 2-Cyanoethylhydrazine
    Assay 95%

    Description

    Applications
    2-Cyanoethylhydrazine is used as pharmaceutical intermediate.

    Solubility
    Soluble in water.

    Notes
    Hygroscopic. Incompatible with oxidizing agents and heat.

    GHS H Statement
    H300-H314
    Fatal if swallowed.
    Causes severe skin burns and eye damage.

    2° to 8°C

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit