Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Cyanoethylhydrazine, 95%

Click to view available options
Quantity:
1g
5g
25g
353-07-1
C3H7N3
85.11
MFCD00043620
VIHRIIARIFUQLC-UHFFFAOYSA-N
2-cyanoethylhydrazine,beta-cyanoethylhydrazine,2-cyanoethyl hydrazine,cyanoethylhydrazine,unii-l6h109c98b,3-hydrazinopropionitrile,propanenitrile, 3-hydrazino,3-hydrazinopropanenitrile
136177
3-hydrazinylpropanenitrile
C(CNN)C#N
353-07-1
C3H7N3
85.11
MFCD00043620
VIHRIIARIFUQLC-UHFFFAOYSA-N
2-cyanoethylhydrazine,beta-cyanoethylhydrazine,2-cyanoethyl hydrazine,cyanoethylhydrazine,unii-l6h109c98b,3-hydrazinopropionitrile,propanenitrile, 3-hydrazino,3-hydrazinopropanenitrile
136177
3-hydrazinylpropanenitrile
C(CNN)C#N
Applications
2-Cyanoethylhydrazine is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents and heat.
Flash Point 109°C (228°F)
Boiling Point 76°C to 79°C (0.5mmHg)
Quantity 1g
Linear Formula NCCH2CH2NHNH2
UN Number UN2927
Beilstein 773834
Formula Weight 85.11
Percent Purity 95%
Chemical Name or Material 2-Cyanoethylhydrazine

GHS H Statement
H300-H314
Fatal if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C