Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Cyanoethylhydrazine, 98%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
Specifications
Boiling Point | 76°C to 79°C (0.5mmHg) |
Density | 1.0530g/mL |
Flash Point | 109°C |
Specific Gravity | 1.053 |
Refractive Index | 1.4740 to 1.476 |
Color | Yellow |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Quantity | 5g |
CAS Min % | 97.5 |
Show More |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove
GHS Signal Word: Danger
TSCA : TSCA
Recommended Storage : product darkens on exposure to air