Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Cyanomethyl-4-fluorobenzeneboronic acid pinacol ester, 96%

Click to view available options
Quantity:
250mg
1g
2-Cyanomethyl-4-fluorophenylboronic acid pinacol ester
2-Cyanomethyl-4-fluorophenylboronic acid pinacol ester
Quantity 1g
Formula Weight 261.1
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 2-Cyanomethyl-4-fluorobenzeneboronic acid pinacol ester

TSCA : No

Recommended Storage : Ambient temperatures