Alfa Aesar™ 2-Cyclopropyl-D-alanine, 95%

Manufacturer: Alfa Aesar™ H5200403

Quantity 1g

 View more versions of this product

Catalog No. AAH5200403

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 121786-39-8
    Molecular Formula C6H11NO2
    Formula Weight 129.16
    MDL Number MFCD02259520
    Synonym (R)-2-Amino-3-cyclopropylpropionic acid
    Chemical Name or Material 2-Cyclopropyl-D-alanine

    Specifications

    Formula Weight 129.16
    Molecular Formula C6H11NO2
    MDL Number MFCD02259520
    CAS 121786-39-8
    Synonym (R)-2-Amino-3-cyclopropylpropionic acid
    Quantity 1g
    Chemical Name or Material 2-Cyclopropyl-D-alanine
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit