Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare 2-D Fractionation Kit

Simplify the analysis of complex protein mixtures. GE Healthcare 2-D Fractionation Kit reduces the amount and number of protein species loaded into the gel matrix. The proven technology resolves complex protein samples into six easily manageable fractions.

Manufacturer:  GE Healthcare 80650104

Catalog No. 45-002-446

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Fractionation makes it possible to isolate groups of proteins, or fractions, from the total proteome. This allows for improved resolution when an individual fraction is analyzed, provides less crowded 2-D maps, simplifies analysis and interpretation, and increases the chances of discovering novel proteins of diagnostic or therapeutic interest.
  • Resolves complex protein samples into six fractions
  • High sample recovery with no loss of protein
  • Produces more abundant spots
  • Enhances the visibility and detection of low-abundance proteins
  • Can be scaled up for larger samples
  • Compatible with other downstream separation techniques, such as 2-D electrophoresis
Specifications

Specifications

1 Kit: 50 mL Lysis Buffer, 15 mL Fraction Precipitant, 2 x 30 mL Precipitant, 2 x 30mL Coprecipitant, 50 mL Wash Buffer
50mL Lysis buffer
15mL Fraction precipitant
2 x 30mL Precipitant
2 x 30mL Coprecipitant
50mL Wash buffer
Lysis buffer: Once opened, store at 2 to 8°C
Fraction precipitant: Store at ambient
Precipitant: Store at ambient
Co-precipitant: Store at ambient
Wash buffer: Place at −20°C for 1 to 2 hours before first use and thereafter store −20°C
Solubilizer: Store powder at ambient
Diluent: Once opened, store at 2 to 8°C
2-D Protein Fractionation Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit