Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Deoxy-D-Ribose, >98%, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
5g
100g
Packaging:
Plastic bottle
533-67-5
C5H10O4
134.131
MFCD00135904
ASJSAQIRZKANQN-CRCLSJGQSA-N
2-deoxy-d-ribose,thyminose,3s,4r-3,4,5-trihydroxypentanal,deoxyribose,2-deoxy-d-erythro-pentose,2-deoxyribose,d-drib,2-deoxy-d-arabinose,unii-lsw4h01241,2-deoxy-d-erythropentose
5460005
CHEBI:28816
(3S,4R)-3,4,5-trihydroxypentanal
C(C=O)C(C(CO)O)O
533-67-5
C5H10O4
134.131
MFCD00135904
ASJSAQIRZKANQN-CRCLSJGQSA-N
2-deoxy-d-ribose,thyminose,3s,4r-3,4,5-trihydroxypentanal,deoxyribose,2-deoxy-d-erythro-pentose,2-deoxyribose,d-drib,2-deoxy-d-arabinose,unii-lsw4h01241,2-deoxy-d-erythropentose
5460005
CHEBI:28816
(3S,4R)-3,4,5-trihydroxypentanal
C(C=O)C(C(CO)O)O
Packaging Plastic bottle
Melting Point 76°C to 85°C
Quantity 5g
Formula Weight 134.13
Physical Form Powder
Percent Purity ≥98%
Chemical Name or Material 2-Deoxy-D-Ribose