Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Deoxyguanosine Monohydrate, Synthetic, >98% (Dry Basis), Thermo Scientific Chemicals

Catalog Number aaj1430409
Click to view available options
Quantity:
10 g
Packaging:
Plastic bottle
312693-72-4
C10H15N5O5
285.26
MFCD00150760
LZSCQUCOIRGCEJ-VDPVQVSMNA-N
2 '-deoxyguanosine monohydrate, 2 '-deoxyguanosine hydrate, 2-deoxyguanosine hydrate, 2-amino-9-2 r,4 s,5 r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1 h-purin-6 9 h-one hydrate, 2-amino-9-2 r,4 s,5 r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1 h-purin-6 9 h-one hydrate 1:x, 2 acute-deoxyguanosine, deoxyguanosine hydrate, 2'deoxyguanosine monohydrate, 2'-deoxyguanosinemonohydrate
16218597
2-amino-9-[(2 R,4 S,5 R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3 H-purin-6-one;hydrate
O.NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1
312693-72-4
C10H15N5O5
285.26
MFCD00150760
LZSCQUCOIRGCEJ-VDPVQVSMNA-N
2 '-deoxyguanosine monohydrate, 2 '-deoxyguanosine hydrate, 2-deoxyguanosine hydrate, 2-amino-9-2 r,4 s,5 r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1 h-purin-6 9 h-one hydrate, 2-amino-9-2 r,4 s,5 r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1 h-purin-6 9 h-one hydrate 1:x, 2 acute-deoxyguanosine, deoxyguanosine hydrate, 2'deoxyguanosine monohydrate, 2'-deoxyguanosinemonohydrate
16218597
2-amino-9-[(2 R,4 S,5 R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3 H-purin-6-one;hydrate
O.NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1
Packaging Plastic bottle
Quantity 10 g
Formula Weight 285.26
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material 2 '-Deoxyguanosine monohydrate
EINECSNumber 213-505-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit