Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Di-p-tolylphosphino)benzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
500mg
1202865-03-9
C21H19OP
318.35
MFCD15143661
IDDRQJVTYBKJLP-UHFFFAOYSA-N
2-di-p-tolylphosphino benzaldehyde,2-bis 4-methylphenyl phosphanyl benzaldehyde,acmc-20aon9,2-bis 4-methylphenyl phosphino benzaldehyde,2-di-p-tolylphosphino benzaldehyde,min
71310894
2-bis(4-methylphenyl)phosphanylbenzaldehyde
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=CC=CC=C3C=O
1202865-03-9
C21H19OP
318.35
MFCD15143661
IDDRQJVTYBKJLP-UHFFFAOYSA-N
2-di-p-tolylphosphino benzaldehyde,2-bis 4-methylphenyl phosphanyl benzaldehyde,acmc-20aon9,2-bis 4-methylphenyl phosphino benzaldehyde,2-di-p-tolylphosphino benzaldehyde,min
71310894
2-bis(4-methylphenyl)phosphanylbenzaldehyde
CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=CC=CC=C3C=O
Quantity 500mg
Color Yellow
Melting Point 109.0°C to 110.5°C
Elemental Analysis 76.05 to 82.40% C
Infrared Spectrum Authentic
NMR (1H-NMR) Consistent with structure, (31P-NMR) Consistent with structure, NMR (19F-NMR), Consistent with structure
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 318.35
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-(Di-p-Tolylphosphino)benzaldehyde
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning