Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Di-tert-butylphosphino)-2′-methylbiphenyl, 98+%, Acros Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
255837-19-5
C21H29P
312.437
MFCD03453047
UJONYAVMBYXBJQ-UHFFFAOYSA-N
di-tert-butyl 2'-methyl-1,1'-biphenyl-2-yl phosphine,2-di-tert-butylphosphino-2'-methylbiphenyl,tbumephos,t-butyl mephos,2-di-t-butylphosphino-2'-methylbiphenyl,phosphine, bis 1,1-dimethylethyl 2'-methyl 1,1'-biphenyl-2-yl,di-tert-butyl 2'-methylbiphenyl-2-yl phosphine,di-tert-butyl 2-2-methylphenyl phenyl phosphane
2734937
ditert-butyl-[2-(2-methylphenyl)phenyl]phosphane
CC1=CC=CC=C1C2=CC=CC=C2P(C(C)(C)C)C(C)(C)C
255837-19-5
C21H29P
312.437
MFCD03453047
UJONYAVMBYXBJQ-UHFFFAOYSA-N
di-tert-butyl 2'-methyl-1,1'-biphenyl-2-yl phosphine,2-di-tert-butylphosphino-2'-methylbiphenyl,tbumephos,t-butyl mephos,2-di-t-butylphosphino-2'-methylbiphenyl,phosphine, bis 1,1-dimethylethyl 2'-methyl 1,1'-biphenyl-2-yl,di-tert-butyl 2'-methylbiphenyl-2-yl phosphine,di-tert-butyl 2-2-methylphenyl phenyl phosphane
2734937
ditert-butyl-[2-(2-methylphenyl)phenyl]phosphane
CC1=CC=CC=C1C2=CC=CC=C2P(C(C)(C)C)C(C)(C)C
Color White
Packaging Glass bottle
Melting Point 91.0°C to 93.0°C
Quantity 1g
Infrared Spectrum Authentic
Formula Weight 312.43
Physical Form Crystalline Powder
Percent Purity ≥98%
Chemical Name or Material 2-(Di-tert-butylphosphino)-2′-methylbiphenyl

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.