Alfa Aesar™ 2-Di-tert-butylphosphino-3,4,5,6-tetramethyl-2',4',6'-triisopropylbiphenyl, 98% min

Manufacturer: Alfa Aesar™ H35719-06

Quantity 5g

 View more versions of this product

Catalog No. AAH3571906

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 857356-94-6
    Molecular Formula C33H53P
    Formula Weight 480.75
    MDL Number MFCD09038436
    Synonym Me4 t-butylXPhos; Tetramethyl di-tBuXPhos
    Chemical Name or Material 2-Di-tert-butylphosphino-3,4,5,6-tetramethyl-2',4',6'-triisopropylbiphenyl

    Specifications

    Formula Weight 480.75
    Molecular Formula C33H53P
    MDL Number MFCD09038436
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 166° to 168°C
    CAS 857356-94-6
    Synonym Me4 t-butylXPhos; Tetramethyl di-tBuXPhos
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    Chemical Name or Material 2-Di-tert-butylphosphino-3,4,5,6-tetramethyl-2',4',6'-triisopropylbiphenyl
    Assay ≥98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit