Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Di-tert-butylphosphino)biphenyl, 99%, ACROS Organics™

Click to view available options
Quantity:
500mg
5g
25g
Packaging:
Glass bottle
224311-51-7
C20H27P
298.41
MFCD01862440
CNXMDTWQWLGCPE-UHFFFAOYSA-N
2-di-tert-butylphosphino biphenyl,johnphos,2-di-t-butylphosphino biphenyl,biphenyl-2-yl-di-tert-butyl-phosphane,2-biphenyl di-tert-butylphosphine,2-biphenylyl di-tert-butylphosphine,2-di-tert-butylphosphino-biphenyl,biphenyl-2-yldi-tert-butylphosphine,di-tert-butyl 2-phenylphenyl phosphane,1,1'-biphenyl-2-yldi-tert-butylphosphine
2734215
ditert-butyl-(2-phenylphenyl)phosphane
CC(C)(C)P(C1=CC=CC=C1C2=CC=CC=C2)C(C)(C)C
224311-51-7
C20H27P
298.41
MFCD01862440
CNXMDTWQWLGCPE-UHFFFAOYSA-N
2-di-tert-butylphosphino biphenyl,johnphos,2-di-t-butylphosphino biphenyl,biphenyl-2-yl-di-tert-butyl-phosphane,2-biphenyl di-tert-butylphosphine,2-biphenylyl di-tert-butylphosphine,2-di-tert-butylphosphino-biphenyl,biphenyl-2-yldi-tert-butylphosphine,di-tert-butyl 2-phenylphenyl phosphane,1,1'-biphenyl-2-yldi-tert-butylphosphine
2734215
ditert-butyl-(2-phenylphenyl)phosphane
CC(C)(C)P(C1=CC=CC=C1C2=CC=CC=C2)C(C)(C)C
Melting Point 84°C to 87°C
Color White
Quantity 500mg
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Merck Index 15, 1469
Formula Weight 298.41
Physical Form Crystals or Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2-(Di-tert-butylphosphino)biphenyl, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

TSCA : TSCA