Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl, 98%, Acros Organics

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
251320-85-1
C27H37P
392.56
MFCD09744023
PQWWLQRSCJJOIS-UHFFFAOYSA-N
2-dicyclohexylphosphino-2'-isopropylbiphenyl,2-dicyclohexylphosphino-2'-i-propylbiphenyl,dicyclohexyl 2'-propan-2-yl 1,1'-biphenyl-2-yl phosphane,dicyclohexyl 2'-isopropyl-1,1'-biphenyl-2-yl phosphane
23420463
dicyclohexyl-[2-(2-propan-2-ylphenyl)phenyl]phosphane
CC(C)C1=CC=CC=C1C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4
251320-85-1
C27H37P
392.56
MFCD09744023
PQWWLQRSCJJOIS-UHFFFAOYSA-N
2-dicyclohexylphosphino-2'-isopropylbiphenyl,2-dicyclohexylphosphino-2'-i-propylbiphenyl,dicyclohexyl 2'-propan-2-yl 1,1'-biphenyl-2-yl phosphane,dicyclohexyl 2'-isopropyl-1,1'-biphenyl-2-yl phosphane
23420463
dicyclohexyl-[2-(2-propan-2-ylphenyl)phenyl]phosphane
CC(C)C1=CC=CC=C1C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4
Infrared Spectrum Authentic
Quantity 1g
Color White
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: insoluble
Formula Weight 392.56
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning