Alfa Aesar™ 2-Dicyclohexylphosphino-2'-methylbiphenyl, 98%

Manufacturer: Alfa Aesar™ H27647-06

Quantity 5g

 View more versions of this product

Catalog No. AAH2764706

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 251320-86-2
    Molecular Formula C25H33P
    Formula Weight 364.51
    MDL Number MFCD03094577
    Chemical Name or Material 2-Dicyclohexylphosphino-2'-methylbiphenyl

    Specifications

    Formula Weight 364.51
    Melting Point 107° to 110°C
    Molecular Formula C25H33P
    MDL Number MFCD03094577
    CAS 251320-86-2
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Quantity 5g
    Chemical Name or Material 2-Dicyclohexylphosphino-2'-methylbiphenyl
    Assay 98%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit