Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dicyclohexylphosphino)-2-methylbiphenyl, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
251320-86-2
C25H33P
364.513
MFCD03094577
GPVWUKXZFDHGMZ-UHFFFAOYSA-N
2-dicyclohexylphosphino-2'-methylbiphenyl,mephos,dicyclohexyl 2'-methyl-1,1'-biphenyl-2-yl phosphine,dicyclohexyl 2-2-methylphenyl phenyl phosphane,dicyclohexyl 2'-methyl-1,1'-biphenyl-2-yl phosphane,phosphine, dicyclohexyl 2'-methyl 1,1'-biphenyl-2-yl,dicyclohexyl-2'-methyl-biphenyl-2-yl-phosphane,dicyclohexyl 2'-methyl 1,1'-biphenyl-2-yl-phosphine
2734938
dicyclohexyl-[2-(2-methylphenyl)phenyl]phosphane
CC1=CC=CC=C1C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4
251320-86-2
C25H33P
364.513
MFCD03094577
GPVWUKXZFDHGMZ-UHFFFAOYSA-N
2-dicyclohexylphosphino-2'-methylbiphenyl,mephos,dicyclohexyl 2'-methyl-1,1'-biphenyl-2-yl phosphine,dicyclohexyl 2-2-methylphenyl phenyl phosphane,dicyclohexyl 2'-methyl-1,1'-biphenyl-2-yl phosphane,phosphine, dicyclohexyl 2'-methyl 1,1'-biphenyl-2-yl,dicyclohexyl-2'-methyl-biphenyl-2-yl-phosphane,dicyclohexyl 2'-methyl 1,1'-biphenyl-2-yl-phosphine
2734938
dicyclohexyl-[2-(2-methylphenyl)phenyl]phosphane
CC1=CC=CC=C1C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4
Melting Point 108°C to 112°C
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 364.51
Physical Form Crystals or Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-(Dicyclohexylphosphino)-2';-methylbiphenyl
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.