Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Dicyclohexylphosphino-2'-(N,N-dimethylamino)biphenyl, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
213697-53-1
C26H36NP
393.555
MFCD02183572
ZEMZPXWZVTUONV-UHFFFAOYSA-N
2-dicyclohexylphosphino-2'-n,n-dimethylamino biphenyl,davephos,2'-dicyclohexylphosphino-n,n-dimethyl-1,1'-biphenyl-2-amine,2'-dicyclohexylphosphino-n,n-dimethylbiphenyl-2-amine,2-dicyclohexylphosphino-2'-dimethylamino biphenyl,unii-9877onu78f,2-2-dicyclohexylphosphanyl phenyl-n,n-dimethylaniline,2'-dicyclohexylphosphanyl-n,n-dimethyl-1,1'-biphenyl-2-amine
2734217
2-(2-dicyclohexylphosphanylphenyl)-N,N-dimethylaniline
CN(C)C1=CC=CC=C1C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4
213697-53-1
C26H36NP
393.555
MFCD02183572
ZEMZPXWZVTUONV-UHFFFAOYSA-N
2-dicyclohexylphosphino-2'-n,n-dimethylamino biphenyl,davephos,2'-dicyclohexylphosphino-n,n-dimethyl-1,1'-biphenyl-2-amine,2'-dicyclohexylphosphino-n,n-dimethylbiphenyl-2-amine,2-dicyclohexylphosphino-2'-dimethylamino biphenyl,unii-9877onu78f,2-2-dicyclohexylphosphanyl phenyl-n,n-dimethylaniline,2'-dicyclohexylphosphanyl-n,n-dimethyl-1,1'-biphenyl-2-amine
2734217
2-(2-dicyclohexylphosphanylphenyl)-N,N-dimethylaniline
CN(C)C1=CC=CC=C1C2=CC=CC=C2P(C3CCCCC3)C4CCCCC4
Melting Point 116°C to 121°C
Quantity 5g
Packaging Glass bottle
Color White to Yellow
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Merck Index 15, 1469
Formula Weight 393.55
Physical Form Crystals or Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Dicyclohexylphosphino-2'-(N, N-dimethylamino)biphenyl, 97%
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

TSCA : TSCA