Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Dicyclohexylphosphino-2'-(N,N-dimethylamino)biphenyl, 98%

Manufacturer:  Alfa Aesar™ H27678-03

Quantity 1g

 View more versions of this product

Catalog No. AAH2767803

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

213697-53-1
393.56
DavePhos; 2-Dimethylamino-2'-dicyclohexylphosphinobiphenyl
C26H36NP
MFCD02183572
2-Dicyclohexylphosphino-2'-(N,N-dimethylamino)biphenyl

Specifications

1g
C26H36NP
MFCD02183572
DavePhos; 2-Dimethylamino-2'-dicyclohexylphosphinobiphenyl
98%
213697-53-1
393.56
121° to 124°C
2-Dicyclohexylphosphino-2'-(N,N-dimethylamino)biphenyl

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit