Learn More
Strem Chemical, Inc.Supplier Diversity Partner 2-(DICYCLOHEXYLPHOSPHINO)-3,6- DFS Item
Manufacturer:  Strem Chemical, Inc.Supplier Diversity Partner 151152.5
2-(Dicyclohexylphosphino)-3,6-dimethoxy-2'-4'-6'-tri-i-propyl-1,1'-biphenyl, min. 98% BrettPhos; Size: 5g; [1070663-78-3]; C35H53O2P; Mol Wt:536.77; Color & Form: white xtl.
Catalog No. NC0133236