Learn More
Accela Chembio IncSupplier Diversity Partner 2-DICYCLOHEXYLPHOSPHINO 5G DFS Item
Manufacturer:  Accela Chembio IncSupplier Diversity Partner SY0098175G
2-DICYCLOHEXYLPHOSPHINO-2',6'-DIISOPROPOXYBIPHENYL, 5G, 787618-22-8, MFCD06798294, 95%, RETEST PERIOD: 540DAYS, LIGHT SENSITIVE/NITROGEN OR ARGON/+4 ℃
Catalog No. 50-113-0866