Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dicyclohexylphosphino)biphenyl, 98%, ACROS Organics™

Click to view available options
Quantity:
500mg
2g
Packaging:
Glass bottle
247940-06-3
C24H31P
350.486
MFCD01862441
LCSNDSFWVKMJCT-UHFFFAOYSA-N
2-dicyclohexylphosphino biphenyl,cyjohnphos,1,1'-biphenyl-2-yldicyclohexylphosphine,2-biphenyl dicyclohexylphosphine,2-biphenylyl dicyclohexylphosphine,biphenyl-2-yldicyclohexylphosphine,dicyclohexyl 2-phenylphenyl phosphane,cyclohexyl johnphos,dicyclohexyl-2-phenylphenyl phosphane,2-dicyclohexylphosphino-biphenyl
2734216
dicyclohexyl-(2-phenylphenyl)phosphane
C1CCC(CC1)P(C2CCCCC2)C3=CC=CC=C3C4=CC=CC=C4
247940-06-3
C24H31P
350.486
MFCD01862441
LCSNDSFWVKMJCT-UHFFFAOYSA-N
2-dicyclohexylphosphino biphenyl,cyjohnphos,1,1'-biphenyl-2-yldicyclohexylphosphine,2-biphenyl dicyclohexylphosphine,2-biphenylyl dicyclohexylphosphine,biphenyl-2-yldicyclohexylphosphine,dicyclohexyl 2-phenylphenyl phosphane,cyclohexyl johnphos,dicyclohexyl-2-phenylphenyl phosphane,2-dicyclohexylphosphino-biphenyl
2734216
dicyclohexyl-(2-phenylphenyl)phosphane
C1CCC(CC1)P(C2CCCCC2)C3=CC=CC=C3C4=CC=CC=C4
Color White
Infrared Spectrum Authentic
Melting Point 102°C to 106°C
Quantity 2g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 350.49
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-(Dicyclohexylphosphino)biphenyl
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.