Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Dicyclohexylphosphinoethyl)trimethylammonium chloride 97%, Acros Organics

Click to view available options
Quantity:
5g
181864-78-8
C17H35ClNP
319.898
MFCD05664248
ADRQXJKZPSRURO-UHFFFAOYSA-M
2-dicyclohexylphosphino ethyl trimethylammonium chloride,2-dicyclohexylphosphino ethyl trimethylammoniumchloride,2-dicyclohexylphosphanyl ethyl trimethylazanium chloride,2-dicyclohexylphosphanylethyl trimethyl azanium,chloride,2-dicyclohexylphosphino-n,n,n-trimethylethanaminium chloride,2-dicyclohexylphosphanyl-n,n,n-trimethylethan-1-aminium chloride
15535162
2-dicyclohexylphosphanylethyl(trimethyl)azanium;chloride
C[N+](C)(C)CCP(C1CCCCC1)C2CCCCC2.[Cl-]
181864-78-8
C17H35ClNP
319.898
MFCD05664248
ADRQXJKZPSRURO-UHFFFAOYSA-M
2-dicyclohexylphosphino ethyl trimethylammonium chloride,2-dicyclohexylphosphino ethyl trimethylammoniumchloride,2-dicyclohexylphosphanyl ethyl trimethylazanium chloride,2-dicyclohexylphosphanylethyl trimethyl azanium,chloride,2-dicyclohexylphosphino-n,n,n-trimethylethanaminium chloride,2-dicyclohexylphosphanyl-n,n,n-trimethylethan-1-aminium chloride
15535162
2-dicyclohexylphosphanylethyl(trimethyl)azanium;chloride
C[N+](C)(C)CCP(C1CCCCC1)C2CCCCC2.[Cl-]
Color White
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (C6H11)2P(CH2)2N(CH3)3Cl
Formula Weight 319.9
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-(Dicyclohexylphosphinoethyl)trimethylammonium chloride
Show More Show Less