Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lidocaine, 97.5%, Acros Organics™

Click to view available options
Quantity:
10g
100g
Packaging:
Glass bottle
137-58-6
C14H22N2O
234.343
MFCD00026733
NNJVILVZKWQKPM-UHFFFAOYSA-N
lidocaine,lignocaine,xylocaine,lidoderm,2-diethylamino-n-2,6-dimethylphenyl acetamide,anestacon,esracaine,duncaine,cappicaine,gravocain
3676
CHEBI:6456
2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
CCN(CC)CC(=O)NC1=C(C=CC=C1C)C
137-58-6
C14H22N2O
234.343
MFCD00026733
NNJVILVZKWQKPM-UHFFFAOYSA-N
lidocaine,lignocaine,xylocaine,lidoderm,2-diethylamino-n-2,6-dimethylphenyl acetamide,anestacon,esracaine,duncaine,cappicaine,gravocain
3676
CHEBI:6456
2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide
CCN(CC)CC(=O)NC1=C(C=CC=C1C)C
Infrared Spectrum Authentic
Quantity 100g
Melting Point 66.0°C to 69.0°C
Boiling Point 182.0°C (4.0 mmHg)
Packaging Glass bottle
Color White to Yellow
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97.5%
Merck Index 15, 5535
Formula Weight 234.34
Physical Form Crystalline Powder
Percent Purity 97.5%
Chemical Name or Material Lidocaine
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning