Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Diethylaminoethanol, Reagent, 98%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
100-37-8
C6H15NO
117.19
BFSVOASYOCHEOV-UHFFFAOYSA-N
2-(diethylamino)ethan-1-ol
CCN(CC)CCO
100-37-8
C6H15NO
117.19
BFSVOASYOCHEOV-UHFFFAOYSA-N
2-(diethylamino)ethan-1-ol
CCN(CC)CCO

D1016, 100-37-8, C6H15NO

Spectrum™ 2-Diethylaminoethanol, Reagent is used as a precursor chemical to procaine. In condensate and steam lines, it is used as a corrosion inhibitor because it neutralizes carbonic acid and scavenging oxygen. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Packaging Amber Glass Bottle
Quantity 4 L
CAS Max % 100%
UN Number UN2686
Percent Purity 98%