Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-(Difluoromethoxy)benzaldehyde, 97%
Click to view available options
Quantity:
5g
Description
Applications
2-(Difluoromethoxy)benzaldehyde is an important raw material used as an intermediate in the preparation of pharmaceuticals, agrochemicals, and dyestuffs. It is also involved in the preparation of 4-(2-difluoromethoxyphenyl)- substituted 1,4-dihydropyridine 3,5-diesters.
Solubility
Miscible with 2-propanol and acetic acid.
Notes
Incompatible with strong oxidizing agents.
2-(Difluoromethoxy)benzaldehyde is an important raw material used as an intermediate in the preparation of pharmaceuticals, agrochemicals, and dyestuffs. It is also involved in the preparation of 4-(2-difluoromethoxyphenyl)- substituted 1,4-dihydropyridine 3,5-diesters.
Solubility
Miscible with 2-propanol and acetic acid.
Notes
Incompatible with strong oxidizing agents.
Specifications
Quantity | 5g |
Boiling Point | 88°C to 90°C (9mmHg) |
Flash Point | 106°C (223°F) |
Refractive Index | 1.497 |
Density | 1.284 |
Sensitivity | Air Sensitive |
Beilstein | 389068 |
Solubility Information | Miscible with 2-propanol and acetic acid. |
Formula Weight | 172.13 |
Percent Purity | 97% |
Show More |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon