Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Difluoromethoxy)benzoic acid, 97%

Click to view available options
Quantity:
1g
5g
97914-59-5
C8H6F2O3
188.13
MFCD00236223
AGDOJFCUKQMLHD-UHFFFAOYSA-N
2-difluoromethoxy benzoic acid,benzoic acid, 2-difluoromethoxy,2-difluoromethoxy-benzoic acid,rarechem al bo 1907,timtec-bb sbb029907,art-chem-bb b006808,benzoic acid,2-difluoromethoxy,2-carboxy-alpha,alpha-difluoroanisole,2-bis fluoranyl methoxy benzoic acid
2063348
2-(difluoromethoxy)benzoic acid
C1=CC=C(C(=C1)C(=O)O)OC(F)F
97914-59-5
C8H6F2O3
188.13
MFCD00236223
AGDOJFCUKQMLHD-UHFFFAOYSA-N
2-difluoromethoxy benzoic acid,benzoic acid, 2-difluoromethoxy,2-difluoromethoxy-benzoic acid,rarechem al bo 1907,timtec-bb sbb029907,art-chem-bb b006808,benzoic acid,2-difluoromethoxy,2-carboxy-alpha,alpha-difluoroanisole,2-bis fluoranyl methoxy benzoic acid
2063348
2-(difluoromethoxy)benzoic acid
C1=CC=C(C(=C1)C(=O)O)OC(F)F
Quantity 5g
Melting Point 98°C to 100°C
Formula Weight 188.13
Percent Purity 97%
Chemical Name or Material 2-(Difluoromethoxy)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning