Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Difluoromethoxy)benzyl alcohol, 97+%

Click to view available options
Quantity:
1g
5g
72768-94-6
C8H8F2O2
174.147
MFCD00236226
JJAUYEZYAXAERE-UHFFFAOYSA-N
2-difluoromethoxy benzyl alcohol,2-difluoromethoxy phenyl methanol,2-difluoromethoxy-phenyl-methanol,2-difluoromethoxy phenyl methan-1-ol,2-difluoromethoxybenzyl alcohol,rarechem al bd 0278,attercop-chm at117237,benzenemethanol,2-difluoromethoxy
2736992
[2-(difluoromethoxy)phenyl]methanol
C1=CC=C(C(=C1)CO)OC(F)F
72768-94-6
C8H8F2O2
174.147
MFCD00236226
JJAUYEZYAXAERE-UHFFFAOYSA-N
2-difluoromethoxy benzyl alcohol,2-difluoromethoxy phenyl methanol,2-difluoromethoxy-phenyl-methanol,2-difluoromethoxy phenyl methan-1-ol,2-difluoromethoxybenzyl alcohol,rarechem al bd 0278,attercop-chm at117237,benzenemethanol,2-difluoromethoxy
2736992
[2-(difluoromethoxy)phenyl]methanol
C1=CC=C(C(=C1)CO)OC(F)F
Quantity 5g
Formula Weight 174.15
Percent Purity ≥97%
Chemical Name or Material 2-(Difluoromethoxy)benzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning